missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Butyl-S FF Columns

Product Code. 11584955 Shop All Cytiva Products
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Diameter (English) Inner:
0.275 in.
0.629 in.
Flow Rate:
< 1 mL/min
5 mL/min
Unit Size:
Pack of 5
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11584955

Brand: Cytiva 17097814

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Designed for purification of relatively strong hydrophilic molecules at low salt concentrations

  • Useful for removal of strong hydrophobic contaminants, such as lipoproteins, lipids, pigments and more from biological samples
  • Used successfully for large-scale purification of recombinant Hepatitis B virus surface antigen (rec-HBsAg) from CHO cells
  • Robust production with high reproducibility

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.629 in.
Diameter (Metric) Inner 16 mm
Product Type HiTrap Butyl-S FF column
Flow Rate 5 mL/min
For Use With (Application) Small-Scale Protein Purification as Well as for Screening of Binding and Elution Condition
Quantity 5 x 5 mL
Ligand Type Butyl-S
Material Polypropylene
Max. Pressure 5 bar
Particle Size 45 to 165 μm
pH Range 3 to 13
Support Type 6% Cross-linked Agarose
Storage Requirements 4°C to 30°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.