missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ Q ImpRes Column

Product Code. 16320906 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16320906 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16320906 Supplier Cytiva Supplier No. 29400462
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepacked with a strong anion exchange chromatography resin for high resolution protein purification

  • HiTrap column prepacked with Capto Q ImpRes resin
  • Designed to offer quality and flexibility in process-scale applications
  • Scaling up allows for high resolution combined with quick flow rates
  • Capto Q ImpRes resin couples a quarternary ammonium (Q) strong anion exchanger to a chemically modified, high-flow agarose matrix
  • Matrix provides greater particle rigidity without compromising pore size to allow for high bed heights and purification of viscous samples at high flow rates
  • Combining high binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale
  • Suitable for screening selectivity, binding and elution conditions, and for small-scale purifications
  • Optimized for intermediate and polishing protein purification
  • Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
TRUSTED_SUSTAINABILITY

Specifications

Description High Resolution Protein Purification
Matrix High-Flow Agarose
Flow Rate <4 mL/min
For Use With (Application) Ion Exchange Chromatography
Content And Storage Store at 4°C to 30°C, in 20% Ethanol
Binding Strength >55 mg BSA/mL medium, > 48 mg β-lactoglobulin/mL medium
Format HiTrap™ Column
Max. Operating Pressure 5 bar (70 psi)
Column Type Pre-packed Column
Diameter (Metric) Inner Column 7 mm
Volume (Metric) 1 mL
pH 2 to 14
Quantity 1 Ea.
Length (Metric) 25 mm
Show More Show Less
compliance-icons
Product Identifier
  • hitrap capto q impres
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.