missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ S ImpAct Column

Product Code. 16390896 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16390896 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16390896 Supplier Cytiva Supplier No. 29400459
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Prepacked with a cation exchange chromatography resin for screening and small-scale protein purifications

  • Prepacked columns for small scale purifications, as well as optimization of methods and parameters, such as sample load and binding conditions
  • HiTrap™ Capto™ S ImpAct columns provide fast, reproducible, and easy separations in a convenient format
  • A BioProcess chromatography resin that is developed for large-scale purification of biopharmaceutical proteins
  • Based on the well-established Capto base matrix, it provides many opportunities for improved productivity and straightforward process development
  • Capto S ImpAct meets industrial demands with security of supply and comprehensive technical and regulatory support
  • Columns are best utilized when they are connected to liquid chromatography systems such as ÄKTA systems
  • High binding capacity, > 100 mg MAb/mL resin
  • Efficient aggregate removal at high load of monoclonal antibodies
  • Flexibility of design thanks to a large operational window of flow rates and bed heights for easy optimization and scaling
TRUSTED_SUSTAINABILITY

Specifications

Description Efficient Aggregate Removal at High Load of Monoclonal Antibodies
Flow Rate <4 mL/min
For Use With (Application) Ion Exchange Chromatography
Ligand Type Sulfoethyl (S)
Content And Storage Store at 4°C to 30°C, 20% Ethanol
Binding Strength >100 mg MAb/mL resin
Format HiTrap™ Column
Max. Operating Pressure 5 bar (70 psi)
Column Type Pre-packed Column
Diameter (Metric) Inner Column 7 mm
Volume (Metric) 1 mL
pH 4 to 12
Quantity 1 Ea.
Length (Metric) 25 mm
Ion Exchange Group Strong Cation
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Capto S ImpAct, 1 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.