missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HiTrap™ Chromatography Columns with MabSelect™ VL Protein L Resin

Product Code. 17841532 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
Bed Volume:
1 mL
5 mL
Diameter (Metric) Inner:
7 mm
16 mm
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
25mL
5mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Bed Volume Diameter (Metric) Inner Flow Rate Unit Size
17841532 1 x 1 mL 1 mL 7 mm <4 mL/min. Each
17847774 5 x 1 mL 1 mL 7 mm <4 mL/min. 5mL
17897764 1 x 5 mL 5 mL 16 mm <20 mL/min. 5mL
17817774 5 x 5 mL 5 mL 16 mm <20 mL/min. 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17841532 Supplier Cytiva™ Supplier No. 17542051
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Protein L affinity chromatography columns for capture of antibody Fabs, bispecifics, and other variants containing a kappa light chain.

  • Redesigned protein L ligand captures antibody Fabs, bispecifics, and other variants containing a kappa light chain with high binding capacity and improved alkaline stability
  • High dynamic binding capacity for efficient capture of target molecules
  • Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
  • Provides good resolution for product-related impurities in the capture of bispecific antibodies

As the diversity of antibody variants increases, so does the need for chromatography resins that can capture and purify these variants. As an alternative to protein A, protein L can be used to capture antibody Fabs, bispecific antibodies, and other molecules containing kappa variants of the variable light (VL) chain. Protein L binds human kappa light chain subtypes 1, 3, and 4 and mouse kappa light chain subtype 1.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7 mm
Quantity 1 x 1 mL
Height (English) 1 in.
Height (Metric) 25 mm
Flow Rate <4 mL/min.
Max. Pressure 5 bar
Column Type Affinity Column
For Use With (Application) Antibody Affinity Chromatography
compliance-icons
Product Identifier
  • HiTrap MabSelect VL, 1 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.