missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Fast Flow IEX Selection Kit

Product Code. 15518501 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15518501 5 x 5 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518501 Supplier Cytiva Supplier No. 90100200
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fast flow IEX selec kit HiTrap

  • HiTrap Fast Flow IEX Selection Kit
  • Storage: 4 to 30°C, 20% Ethanol
  • Supplied in packs of 5 x 5mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C to 30°C. 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 5 mL
Quantity 5 x 5 mL
compliance-icons
Product Identifier
  • Fast Flow IEX select kit HiTrap
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.