missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ GCSFSelect

Prepacked columns specifically designed for the purification of recombinant Granulocyte-colony stimulating factor (G-CSF). Based on a highly rigid agarose base matrix that allows for high flow rates at large production scales.

Brand:  Cytiva 17548311

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 15539271

  • 1030.00 € / Pack of 5
Estimated Shipment: 08-07-2024
to see stock.
Add to basket

Description

Description

  • Granulocyte-colony stimulating factor (G-CSF) is a hormone that stimulates production of white blood cells in bone marrow
  • For a highly selective purification step, the affinity ligand is based on a single-chain antibody fragment directed against G-CSF
  • To facilitate binding of the target molecule, the ligand is attached to the base matrix through a hydrophilic spacer arm
  • Ligand is produced in a yeast expression system where fermentation, subsequent purification, and formulation are performed in the absence of animal-derived components
  • GCSFSelect's rigid agarose base matrix allows for processing of large sample volumes
  • Composition: Ligand coupled to Capto via stable amide bonds
Specifications

Specifications

4 mL/min
Single-Chain Antibody Fragment Directed Against G-CSF
2 to 12
3.9 mg/column
Column
Purification of Recombinant Granulocyte-Colony Stimulating Factor (G-CSF)
75 μm
5/Pk.
1 mL
Granulocyte-Colony Stimulating Factor (G-CSF)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers