missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ IEX HP Prepacked Columns

Product Code. 11748508 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
Quantity:
5 Pk.
Each
Flow Rate:
<20 mL/min.
<4 mL/min.
Media Type:
Q Sepharose HP
SP Sepharose HP
Unit Size:
1mL
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity Flow Rate Media Type unitSize
11748508 5 mL 5 Pk. <20 mL/min. SP Sepharose HP Pack of 5
10647265 1 mL 5 Pk. <4 mL/min. SP Sepharose HP Pack of 5
11758508 1 mL 5 Pk. <4 mL/min. Q Sepharose HP Pack of 5
11768508 5 mL 5 Pk. <20 mL/min. Q Sepharose HP Pack of 5
15157956 1 mL Each <4 mL/min. SP Sepharose HP 1mL
15177956 1 mL Each <4 mL/min. Q Sepharose HP 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11748508 Supplier Cytiva Supplier No. 17115201
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Use for high-resolution, small-scale protein purification. Cytiva Life Sciences™ HiTrap™ IEX HP Prepacked Columns are prepacked, ready-to-use Sepharose™ High Performance strong cation exchange columns.

 

  • Prepacked with Q or SP Sepharose High Performance
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatography system
  • Pressure Max: 3 bar
  • 1 and 5mL polypropylene columns
Media:
  • Particle size: 24 to 44μm
  • pH stability Working Range: 4 to 13

 

Q Sepharose (45-000-193, -194, 515)

  • Ligand: Quaternary amine
  • Ion Exchanger Type: Strong anion exchanger
  • Ionic Capacity: 0.14 to 0.20mmol Cl-/mL

SP Sepharose (45-000-191, -192, -514) 

  • Ligand: Sulphopropyl
  • Ion Exchanger Type: Strong cation exchanger
  • Ionic Capacity: 0.15 to 0.20mmol H+/mL
1mL columns
  • Bed Dimensions: 7 x 25mm
  • Flow velocity: max 4mL/min.

5mL columns
  • Bed Dimensions: 16 x 25mm
  • Flow velocity: max 20mL/min.

 

TRUSTED_SUSTAINABILITY

Specifications

Description HiTrap SP HP, 5 x 5 mL
Diameter (English) Inner Column 0.6 in.
Flow Rate <20 mL/min.
For Use With (Application) HiTrap SP HP is a strong cation exchange chromatography column for high-resolution, small-scale protein purification.
Media Type SP Sepharose HP
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
For Use With (Equipment) Designed for use with syringe, peristaltic pump or chromatography system
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 5 mL
Quantity 5 Pk.
compliance-icons
Product Identifier
  • HiTrap SP 5 ml, 5 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.