missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Complete instructions for use and method optimization, recommendations for sample preparation, and a variety of connectors for syringe, peristaltic pump, and chromatography system
- Prepacked with a thiophilic adsorption medium, 2-mercaptopyridine coupled to Sepharose™ High Performance medium
- Recovery of IgM is high, with excellent activity retention; typical purity, >80%
- Purification of human IgM possible; binding capacity: 5mg human IgM/mL medium
- Designed for use with a syringe, peristaltic pump, or chromatography system
Order Info
Supplied in packs of five 1mL columns
Specifications
Specifications
| Bed Volume | 1 mL |
| Binding Capacity | 5 mg Human IgM/mL Medium |
| Diameter (English) Inner | 0.62 in. |
| Diameter (Metric) Inner | 1.6 cm |
| Description | HiTrap IgM Purification HP is a 1 mL column prepacked with Sepharose High Performance for fast and efficient purification of monoclonal IgM. |
| Flow Rate | <4 mL/min. |
| Content And Storage | 4 to 8°C, 20% Ethanol |
| Quantity | 5 x 1 mL |
| For Use With (Application) | Chromatography System, Manual |
| Type | Pre-packed Column |
| Show More |
Safety and Handling
Product Identifier
- HiTrap IgM Purification 1 ml, 5 x 1 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?