missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- IgSelect is an affinity resin designed for the purification of human IgG from different sources.
- The ligand, a 14 kD single chain antibody fragment has been designed for excellent selectivity towards Fc domain of human IgG, enabling high purity in one step.
- The IgSelect has affinity for all subclasses of human IgG, making it an excellent complement to protein A based MabSelect media, which has no affinity for IgG3.
- The ability to bind all subclasses of IgG makes IgSelect an excellent choice for purification of polyclonal human IgG, such as in plasma applications
Specifications
Specifications
| Bed Volume | 1 mL |
| Diameter (Metric) Inner | 7 mm |
| Flow Rate | <600 cm/hr. |
| Content And Storage | 4 to 8 °C, 20% Ethanol |
| Quantity | 5 x 1 mL |
| For Use With (Application) | For purification of polyclonal human IgG, transgenic IgG, and monoclonal IgG3, IgG4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?