missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ IMAC Columns
Shop All Cytiva Products
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Diameter (English) Inner:
0.27 in.
0.62 in.
Particle Size:
34 μm
90 μm
Unit Size:
Pack of 5
Description
Includes
Instructions, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems
- Conveniently charge with your metal of choice
- Can be used with a syringe, peristaltic pump, or liquid chromatography system
- Available prepacked with IMAC Sepharose™ High Performance (HP), with small bead size for high performance purification, or IMAC Sepharose 6 Fast Flow (FF), with larger bead size for easy scaleup
- High binding capacity: ≥40mg histidine-taged protein/mL medium (protein/metal ion dependent)
- Stable in the presence of all commonly used buffers
- Available in 1mL and 5mL sizes
Spécification
Spécification
| Bed Volume | 1 mL |
| Diameter (English) Inner | 0.27 in. |
| Flow Rate | <4 mL/min. |
| Diameter (Metric) Inner | 7 mm |
| Kit Contents | IMAC Sepharose |
| For Use With (Equipment) | Syringe, peristaltic pump or liquid chromato graphy system |
| Material | Polypropylene |
| Max. Pressure | 42 psi |
| pH Range | 3 to 12 |
| Particle Size | 34 μm |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- HiTrap IMAC HP, 1 ml, 5 x 1 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu