missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap KappaSelect Media

Product Code. 14811611 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bed Volume (Metric) unitSize
14811611 1 mL Pack of 5
11380212 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 14811611 Supplier Cytiva Supplier No. 17545811

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 5 x 1 mL
Stability Stable in all commonly used aqueous buffers
Storage Requirements 4 to 8 deg.C, 20% Ethanol
Binding Capacity 15 mg Fab/mL of medium2)
Product Type HiTrap KappaSelect Media
Particle Size 75 μm
Material Polypropylene
Max. Pressure 300 kPa
Matrix Highly Cross-linked Agarose
pH Range 3 to 10
Bed Volume (Metric) 1 mL
Ligand Type Recombinant protein (Mr 13 000)
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap 5 x 1 ml, KappaSelect
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.