missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ MabSelect™ Prepacked Columns

Product Code. 11545135 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
5 x 5 mL
Product Line:
MabSelect
MabSelect™ Xtra
Unit Size:
Each
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Line Unit Size
11545135 5 x 5 mL MabSelect Pack of 5
11783319 1 x 5 mL MabSelect Each
10106234 5 x 1 mL MabSelect™ Xtra Pack of 5
11550634 5 x 5 mL MabSelect™ Xtra Pack of 5
10359534 5 x 1 mL MabSelect Pack of 5
11793319 1 x 5 mL MabSelect™ Xtra Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11545135 Supplier Cytiva Supplier No. 28408256
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these columns in small-scale MAb purification in a continuous bioprocess. Cytiva Life Sciences™ HiTrap™ MabSelect™ Prepacked Columns for fast purification of monoclonal antibodies (Mabs) from large sample volumes

Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.

  • Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
  • High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system

MabSelect

  • Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
  • Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
MabSelect Xtra™
  • Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
  • Allows binding from samples with high expression levels of the monoclonal antibody
  • Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
MabSelect SuRe™ (Superior Resistance)
  • Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
  • Permits significant cost savings and extension of cycles in regular production

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Pre-packed Columns
Product Line MabSelect
Binding Capacity ∼30 mg/mL Human lgG at 2.4 min. residence time
Diameter (Metric) Inner Column 1.6 cm
Diameter (English) Inner Column 0.62 in.
Flow Rate <20 mL/min.
Volume (Metric) Bed 5 mL
For Use With (Application) For fast purification of monoclonal antibodies from large sample volumes, Protein Purification
Description HiTrap MabSelect columns are prepacked with MabSelect, a BioProcess resin for capturing of mAbs from large sample volumes.
Sample Volume 5 mL
Storage Buffer 20% Ethanol
Content And Storage 4°C to 30°C
Quantity 5 x 5 mL
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • HiTrap MabSelect, 5 ml, 5 x 5 ml
Palabra de advertencia
  • Warning
Clasificación
  • Flammable liquid Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.