missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
Specifications
| Bed Volume | 1 mL |
| Flow Rate | <4 mL/min. |
| Max. Operating Pressure | 5 bar (0.5 MPa, 70 psi) |
| Quantity | 5 x 1 mL |
| For Use With (Application) | Antibody affinity chromatography |
| Type | Pre-packed Column |
Safety and Handling
Product Identifier
- HiTrap MabSelect PrismA
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?