missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap MabSelect SuRe PCC Column

Product Code. 15881838 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
Unit Size:
Each
Product Code. Quantity unitSize
15881838 1 x 5 mL Each
1 options
This item is not returnable. View return policy

Product Code. 15881838

Brand: Cytiva 17549112

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Prepacked with MabSelect SuRe pcc protein A resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC).

  • Convenient prepacked column for fast, reproducible, and easy small-scale purifications.
  • High binding capacity, ∼ 60 mg human IgG/mL resin at 2.4 min residence time
  • High productivity through capture of large mass of monoclonal antibodies in a short period of time.
  • Highly concentrated elution pools for operating flexibility and small unit operations.
  • Allows cost-effective cleaning with 0.1–0.5 M NaOH over hundreds of purification cycles.
  • High protein ligand stability reduces ligand leakage.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner Column 16 mm
Flow Rate <20mL/min.
Volume (Metric) Bed 5 mL
For Use With (Application) Antibody affinity chromatography
Description 16 x 25 mm
Content And Storage 2°C to 8°C, 20% Ethanol
Quantity 1 x 5 mL
compliance-icons
Product Identifier
  • HiTrap MabSelect SuRe pcc 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.