missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.
- Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
- High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
- Designed for use with a syringe, a peristaltic pump, or a chromatography system
MabSelect
- Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
- Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
- Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
- Allows binding from samples with high expression levels of the monoclonal antibody
- Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
- Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
- Permits significant cost savings and extension of cycles in regular production
Specifications
Specifications
| Bed Volume | 5 mL |
| Diameter (English) Inner | 0.62 in. |
| Diameter (Metric) Inner | 1.6 cm |
| Flow Rate | <20 mL/min. |
| Content And Storage | 4°C to 30°C, 20% Ethanol |
| Quantity | 5 Pk. |
| For Use With (Application) | Chromatography System, Manual |
| Type | Pre-packed Column |
| Sample Volume | 5 mL |
| Binding Capacity | ≈35mg Human lgG/mL at 2.4 min. residence time |
Safety and Handling
Product Identifier
- HiTrap MabSelect SuRe
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P405-Store locked up.
- MIXTURE LIST-Contain: ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?