missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ MabSelect™ SuRe Prepacked Columns
Shop All Cytiva Products
Click to view available options
Bed Volume:
1 mL
5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
1mL
Each
Pack of 5
Description
Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.
- Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
- High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
- Designed for use with a syringe, a peristaltic pump, or a chromatography system
MabSelect
- Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
- Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
- Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
- Allows binding from samples with high expression levels of the monoclonal antibody
- Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
- Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
- Permits significant cost savings and extension of cycles in regular production
Specifications
Specifications
| Bed Volume | 5 mL |
| Binding Capacity | ≈35mg Human lgG/mL at 2.4 min. residence time |
| Diameter (English) Inner | 0.62 in. |
| Diameter (Metric) Inner | 1.6 cm |
| Flow Rate | <20 mL/min. |
| Content And Storage | 4°C to 30°C, 20% Ethanol |
| Quantity | Each |
| For Use With (Application) | Chromatography System, Manual |
| Type | Pre-packed Column |
| Sample Volume | 5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction