missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein A HP Prepacked Columns

Product Code. 10676315 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
2 x 1 mL
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
1mL
Each
Pack of 2
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate unitSize
10676315 5 x 1 mL <4 mL/min. Pack of 5
11768438 5 x 5 mL <20 mL/min. Pack of 5
10146184 2 x 1 mL <4 mL/min. Pack of 2
11758438 1 x 5 mL <20 mL/min. Each
15157946 1 x 1 mL <4 mL/min. 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10676315 Supplier Cytiva Supplier No. 17040201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants. Cytiva Life Sciences™ HiTrap™ Protein A HP Prepacked Columns are specially designed, together with the medium to provide fast, simple and easy separations in a convenient format.

  • Prepacked with Protein A Sepharose High Performance media for purification and fractionation of IgG subclasses
  • For use with syringe, pump or chromatography system such as KTA design or FPLC System
  • Binds 20mg human IgG/mL medium
  • Stable over pH 3 to 9 for use and 2 to 10 for cleaning
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • 1 and 5mL columns

Media
  • Ligand: Native protein A
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 20 mg/mL IgG Medium
Flow Rate <4 mL/min.
For Use With (Application) Chromatography System, Manual
For Use With (Equipment) For syringe, peristaltic pump or a chromatography system
Content And Storage 4°C to 8°C, 20% Ethanol
Quantity 5 x 1 mL
compliance-icons
Product Identifier
  • HiTrap Protein A 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.