missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ SP Sepharose Fast Flow IEX Columns

Product Code. 11545025 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
Pack of 5
Product Code. Volume (Metric) unitSize
11545025 5 mL Pack of 5
10723007 1 mL Pack of 5
2 options
This item is not returnable. View return policy

Product Code. 11545025

Brand: Cytiva 17515701

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use for small-scale protein purifications Cytiva Life Sciences™ HiTrap™ SP Sepharose Fast Flow IEX Columns are strong cation exchanger columns for rapid separations either alone or connected in series.

  • Convenient for rapid separations either alone or connected in series
  • Designed for use with syringe, peristaltic pump or chromatographic system
  • High flowrates and good scale-up potential

TRUSTED_SUSTAINABILITY

Specifications

Description HiTrap SP FF
Diameter (English) Inner Column 0.6 in.
Flow Rate <20 mL/min.
For Use With (Application) HiTrap SP FF is prepacked with SP Sepharose Fast Flow, and is a strong cation exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
Content And Storage 4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
Diameter (Metric) Inner Column 16 mm
Volume (Metric) 5 mL
Quantity 5 x 5 mL
compliance-icons
Product Identifier
  • HiTrap SP FF 5 ml, 5 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.