missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals HIV Blot 2.2 (CE)

Product Code. 30694943 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
18 Tests/Kit
36 Tests/Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30694943 36 Tests/Kit 1 set
30694940 18 Tests/Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30694943 Supplier MP Biomedicals Supplier No. 11030036
  • ONLINE EXCLUSIVE PRICE
    1011.54 €
    963.37 € / 1 sets
    Save 48.17 €
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

The Blot is a qualitative enzyme immunoassay for in vitro detection of antibodies to HIV-1 and HIV-2 in human serum or plasma. It can be used as a more specific supplemental test on human serum or plasma specimens found repeatedly reactive using screening procedures such as ELISA.

  • The nitrocellulose strips are precoated with antigenic proteins from partially purified inactivated HIV-1, and a specific HIV-2 synthetic peptide on the same strips.
  • Individual nitrocellulose strips are incubated with diluted serum or plasma and controls.
  • Specific antibodies to HIV-1 and HIV-2, if present in the specimens, will bind to the HIV-1 proteins and HIV-2 peptide on the strips.
  • The strips are washed to remove unbound materials.
  • Antibodies that bind specifically to HIV proteins can be visualized using a series of reactions with goat anti-human IgG conjugated with alkaline phosphatase and the substrate BCIP/NBT.
  • Each strip also includes an internal sample addition control to minimize the risk of false negatives due to operational errors and to ensure the addition of samples.
  • CE-marked.
  • For professional use only.
  • Human serum and plasma.

Specifications

Product Type Western Blot Kit
Detection Method Colorimetric
For Use With (Application) Immunoblot (Western)
Membrane Compatibility Nitrocellulose
Quantity 36 Tests/Kit
Substrate Alkaline Phosphatase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.