missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HM Janelia Fluor 526, SE

Product Code. 18005332
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005332 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18005332 Supplier Tocris Bioscience™ Supplier No. 7312/1
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorogenic spontaneously blinking yellow-emitting dye (Reactive group: NHS ester). Application: dSTORM, SmLSM, STED, SIM and confocal microscopy

Key information: HM Janelia Fluor™ 526, SE is a fluorogenic spontaneously blinking yellow-emitting dye; supplied with an NHS ester reactive group for the labeling of primary amines; hydroxymethyl derivative of JF526. Suitable for live cell imaging. Application: Suitable for confocal microscopy, super-resolution microscopy (SRM) including dSTORM, STED, SIM and single-molecule localization spectroscopy (SmLSM). HM Janelia Fluor™ 526, SE is cell permeable. Properties and Photophysical Data: The spontaneous blinking properties of HM Janelia Fluor™ 526, SE avoids the requirement for strongly reducing dSTORM buffers. NHS ester can be converted to relevant substrate for use in self-labeling tag systems, e.g. HaloTag™ and SNAP-tag™. Excitation and emission maxima (λ) are 526 nm and 550 nm, respectively. To measure the absorbance spectrum of this dye we recommend the following solvent: TFE plus 0.1% TFA. We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals. HaloTag is a trademark of Promega Corporation, and SNAP-tag is a trademark of New England BioLabs, Inc.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dioxopyrrolidin-1-yl 3,6-bis(3,3-difluoroazetidin-1-yl)-2,7-difluoro-3H-spiro[isobenzofuran-1,9-xanthene]-6-carboxylate
CAS 2376841-30-2
Quantity 1 mg
Target Amine-Reactive Janelia Fluor Dyes
Molecular Formula C31H21F6N3O6
Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.