missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hoechst 33342
GREENER_CHOICE

Product Code. 30002028
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002028 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30002028 Supplier Tocris Bioscience™ Supplier No. 5117/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent dye for staining DNA

Cell permeable fluorescent DNA stain; binds minor groove of AT-rich regions. Used to quantify DNA in viable cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 875756-97-1
Quantity 50 mg
Molecular Formula C27H31Cl3N6O
Formula Weight 561.93
Physical Form Solid
Color Yellow
Purity >98%
Molecular Weight (g/mol) 561.94
Chemical Name or Material Hoechst 33342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.