missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
Specifications
| For Use With (Equipment) | SE 250, SE 260, MiniVE™ Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 9 |
| Thickness (English) | 0.03 in. |
| Thickness (Metric) | 0.1 cm |
| Volume (Metric) Well | 5.8μL/ mm |
| Width (Metric) | 0.58 cm |
| Depth (English) | 0.51 in. |
| Depth (Metric) | 1.3 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?