missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Buffer chamber, lid with cables, UV-transparent gel running tray, gel casting tray, 4 foam gaskets, bubble level. Complete system (03-500-130) also includes 1.5mm thick 8-well comb and comb back.
Designed for very fast separations of DNA restriction fragments in agarose gels
- The clear molded base acts as a heat-sink—simply fill the base with coolant and seal
- Cooling the unit prior to a run allows higher voltages to be applied without generating band distortions or damaging the gel and unit
- No taping required for casting
- Electrical safety locks in lid
Specifications
Specifications
| Certifications/Compliance | EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE |
| Gel Size | Mini |
| Max. Current | 500 mA |
| Type | Mini Horizontal Agarose Electrophoresis Unit |
| Humidity Range | Up to 80% |
| Includes | Buffer chamber assembly, safety lid with high voltage Leads, running tray, casting tray, Foam gaskets, bubble level, 1.5mm thick, 8-well comb, comb back |
| Model | Hoefer HE 33 Complete |
| Sample Size | Up to 32 |
| Temperature (English) Maximum | 122°F |
| Temperature (Metric) Maximum | 50°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?