missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems

Product Code. 15192345
Change view
Click to view available options
Includes:
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.07 cm), well locating decals, gel seal
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.15 cm), well locating decals, gel seal
Unit, 0.75 mm Accessories
Unit, 1 mm Accessories
Unit, 1.5 mm Accessories
Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, four spring clamps, gel seal, well locating decal and spacer mate
Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, two 10 x 8cm notched Alumina plates, four spring clamps, gel seal, well locating decal and spacer mate
Height (Metric):
16 cm
18 cm
Length (Metric) Gel:
8 cm
8/10.5 cm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Height (Metric) Length (Metric) Gel Unit Size
15192345 Unit, 1.5 mm Accessories 18 cm 8 cm Each
11925358 Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, two 10 x 8cm notched Alumina plates, four spring clamps, gel seal, well locating decal and spacer mate 16 cm 8 cm Each
11935358 Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, four spring clamps, gel seal, well locating decal and spacer mate 18 cm 8/10.5 cm Each
10137294 Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.07 cm), well locating decals, gel seal 18 cm 8/10.5 cm Each
15182345 Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers (0.15 cm), well locating decals, gel seal 18 cm 8/10.5 cm Each
11519683 Unit, 0.75 mm Accessories 16 cm 8 cm Each
13409436 Unit, 1 mm Accessories 18 cm 8 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15192345 Supplier Hoefer™ Supplier No. SE25010A1.5
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

SE250 Basic System includes: upper buffer chamber, lower buffer chamber, interlocking safety lid with leads, ten 10 x 8cm glass plates, two 10 x 8cm notched alumina plates, four spring clamps, gel seal, well locating decal, and Spacer Mate. SE250 Complete System also includes: Dual Gel Caster, two 0.75mm thick 10-well combs, and two pairs 0.75mm thick spacers. SE260 Basic System includes same items as SE250 Basic Systems, except the glass and alumina plates.SE260 Complete Systems add Dual Gel Caster, ten 10.5 x 10cm glass plates, five 10.5 x 10cm notched alumina plates, and two 10-well combs and two pairs spacers (either 0.75mm or 1.5mm thick; see ordering table).

Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

  • Run two gels at once
  • Produce high-resolution separations in as little as 45 minutes with no band “smiling”
  • Use alumina plates (included with specified models) for efficient cooling
  • Buffer volume: 300 mL

Applications:

Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis

Certifications

CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.

Specifications

Certifications/Compliance EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Type Electrophoresis Unit
Includes Unit, 1.5 mm Accessories
Length (Metric) Gel 8 cm
Height (English) 7 in.
Height (Metric) 18 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.