missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ TE70X and TE77X Semidry Blotting Systems

Product Code. 11905348
Change view
Click to view available options
Type:
Semi Dry Electroblotting System
Includes:
21 x 26 cm Blot Paper
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Power Supply, 14 x 16 cm Blot Paper
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Includes Unit Size
11905348 Semi Dry Electroblotting System Power Supply, 14 x 16 cm Blot Paper Each
11915348 Semi Dry Electroblotting System Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ Each
11995338 Semi Dry Electroblotting System 21 x 26 cm Blot Paper Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11905348 Supplier Hoefer™ Supplier No. TE70XP
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Molded base with iridium oxide anode, top assembly with stainless-steel cathode, power cables, 25 sheets of blotting paper, 50 sheets of cellophane (Mfr. Nos. TE70X and TE70XP only), two solid Mylar™ masks

Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour

  • Transfer up to two polyacrylamide gels in a stack
  • Use minimal buffer
  • Do not generate the excessive heat that can distort bands or denature protein
  • Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
  • Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
  • Available with built-in power supply to save space and simplify transfer

Certifications

CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1

Specifications

Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Content And Storage Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (14 x 16cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
For Use With (Equipment) Electrophoresis Blotting System
Type Semi Dry Electroblotting System
Humidity Range Up to 80%
Includes Power Supply, 14 x 16 cm Blot Paper
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Dimensions (L x W x H) 18.89 x 15 x 4 in. (48 x 38 x 10 cm)
Length (English) 18.89 in.
Length (Metric) 48 cm
Width (English) 15 in.
Width (Metric) 38 cm
Height (English) 4 in.
Height (Metric) 10 cm
Voltage 30 V
Wattage 15 w
Amperage 500 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.