Learn More
Jaytec Glass™ Hoffman Voltameter Glass Unit
Product Code. 12391429
Change view
Click to view available options
Product Type:
Graduated Hoffman Voltameter
Ungraduated Hoffman Voltameter
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12391429 Graduated Hoffman Voltameter Each
12381429 Ungraduated Hoffman Voltameter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12391429 Supplier Jaytec Glass™ Supplier No. EKY710L
  • ONLINE EXCLUSIVE PRICE
    187.00 €
    153.52 € / Pack of 1
    Save 33.48 €
    17% Off
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Hoffman Voltameter Glass Unit is for classroom demonstration of electrolysis to determine the chemical composition of water.
  • Manufactured in borosilicate glass
  • Available Graduated or Ungraduated
  • H-pattern style
  • 50 mL capacity
Specifications
For Use With (Application) To demonstrate electrolysis of water into oxygen and hydrogen
Quantity 0 to 50 mL x 0.2 mL
Product Type Graduated Hoffman Voltameter