missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ HOFFMANN™ Pinch Cock Tube Clamps

Product Code. 11895091
Change view
Click to view available options
Width (Metric):
12 mm
17 mm
20 mm
25 mm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
11895091 20 mm Pack of 10
11875091 12 mm Pack of 10
11885091 17 mm Pack of 10
11805101 25 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11895091 Supplier Bochem™ Supplier No. 12532
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Pinch cock tube clamps

  • Pinch cock type
  • Includes flap

Specifications

Quantity 10/Pk.
Type Pinchcock Tube Clamp
Material Martensitic Steel
Size (Metric) Grip For pipes up to O.D. 40 mm
Width (Metric) 20 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.