Learn More
Thermo Scientific™ Shandon™ Holder dispense hopper Thermo Scientific Shandon
Product Code. 12658136 Shop All Epredia Products
Change view
Click to view available options
\
Product Code.
12658136 \
1 options
Product Code. 12658136 Supplier Thermo Scientific™ Shandon™ Supplier No. A78210002
Quantity Price
1 431.00 € / Pack of 1
6 401.99 € / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Included with unit: CD-ROM with unlicenced demo copy of Thermo Scientific Shandon MicroWriter™ software; Dispenser hoppers – six included; Pre-filled cassette tube sample pack; MicroWriter™ specific RS232 cable; Power cord; Operator manual.
Uses foil tape and a heated stylus to imprint characters that maintain legibility even through processing solutions
  • Made to withstand the workload of busy laboratories, marking approximately 20,000 characters per roll of tape
  • Printing can be inverted for reading in either direction
  • Change cassette colour and style easily with fully automated dispensing and labelling process
  • Easy to change hoppers provide the flexibility of allowing an unlimited number of colours and cassettes
  • Imprints, letters, numbers, Roman numerals and special characters on the cassettes in four font sizes
  • Ability to print barcodes on cassettes
  • Prints on all 45° flat-faced cassettes
  • Choice of collection systems makes organising cassettes a seamless operation in any laboratory environment
Carousel-type cassette marking system that permanently marks tissue cassettes with easy to read black type.