missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Holder Kit Thermometer

Product Code. 17678975 Shop All Ohaus Products
Change view
Click to view available options
Model:
e-G31
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17678975 e-G31 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17678975 Supplier OHAUS™ Supplier No. 30692826
Add to basket
Add to basket
This item is not returnable. View return policy

Holder Kit Thermometer e-G31 is for use with OHAUS™ Guardian 3000™ Hotplate Stirrers.

  • Compatible Models:
    • Hotplate-Stirrer e-G31HS04C
    • Hotplate-Stirrer e-G31HS07C
    • Hotplate-Stirrer e-G31HSRDS
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
For Use With (Equipment) OHAUS Guardian 3000™ Hotplate Stirrers
Model e-G31
Type Thermometer Stand Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.