Learn More
OHAUS™ Holder Probe Kit 1
GREENER_CHOICE
Product Code. 30182889 Shop All Ohaus Products
Change view
Click to view available options
Model:
Holder Probe Kit-1
Unit Size:
1 unit
Product Code. Model unitSize
30182889 Holder Probe Kit-1 1 unit
1 options
Product Code. 30182889
Brand: OHAUS™ 30973298
Only null left
Add to basket
Add to basket
High Quality Accessories for Hot Plate Stirrers
  • Accessory for Guardian™ Hotplates and Stirrers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For Guardian 5000 range
Model Holder Probe Kit-1
Quantity 1
Type Hot Plate Kit
Material Plastic, Stainless Steel