Learn More
DWK Life Sciences Holding Device for Reaction Vessels, from chrome-nickel steel
Product Code. 10675401 Shop All DWK Life Sciences Products
Change view
Click to view available options
Description:
DN 100
DN 150
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10675401 DN 150 Each
10665401 DN 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10675401 Supplier DWK Life Sciences Supplier No. 290735701
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The holding device is attached to a tripod linkage and serves to suspend and hold reaction vessels within the fume hood.
Reaction vessels can be operated and tempered in various ways. To prevent the reaction apparatus from moving during the test, the reaction vessel should ideally be suspended from a tripod linkage by means of a holding device. Cooling and heating media can thus be fed flexibly from below to the reaction vessel and removed again.
  • For fixed suspension of the lid and the reaction vessel on a tripod rod
Specifications
Description DN 150
Material Chrome-Nickel Steel
For Use With (Application) Holder for lids and reaction vessels; Mounting
No. per Pack 1/Pk.
Autoclavable Not Autoclavable