missing translation for 'onlineSavingsMsg'
Learn More

Brady™ GlobalMark™ Hole Punch for Tags

Product Code. 11589153
Click to view available options
For Use With (Equipment):
BBP31, BBP33, BBP85, Global Mark, Powermark Printer
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11589153

Brand: Brady™ 020591

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

GlobalMark™ Hole Punch for Tags for use with Global Mark and Powermark Printers.

  • For Use With: BBP31, BBP33, BBP85, Global Mark, Powermark Printers

Specifications

Product Type Hole Punch
For Use With (Equipment) BBP31, BBP33, BBP85, Global Mark, Powermark Printer
Description Hole Punch for tags
Format Hole Punch
Shape Circle
For Use With (Application) Inspection tagging, Lockout tagout
Quantity 1 Ea.
Alternate Part Number Hole Punch
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.