missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) nitrate pentahydrate, REacton™, 99.9% (REO)

Product Code. 11322538
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11322538

Brand: Thermo Scientific Alfa Aesar 014588.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14483-18-2 | H10HoN3O14 | 441.017 g/mol

Holmium(III) nitrate pentahydrate is used to prepare holmium-beta-cyclodextrin complex by reacting with beta- cyclodextrin, which posses potential pharmacological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14483-18-2
Molecular Formula H10HoN3O14
Molecular Weight (g/mol) 441.017
MDL Number MFCD00149737
InChI Key BJUGWWDCFYEYOA-UHFFFAOYSA-N
Synonym holmium nitrate pentahydrate, ho.3no3.5h2o, holmium iii nitrate hydrate, holmium nitrate, holmium nitrate-water 1/3/5, holmium 3+ pentahydrate trinitronate, holmium iii nitrate pentahydrate, holmium 3+ pentahydrate trinitrate, nitric acid,holmium 3+ salt, pentahydrate 8ci,9ci, holmium iii nitrate pentahydrate trace metals basis
PubChem CID 25021754
IUPAC Name holmium(3+);trinitrate;pentahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Ho+3]

Specifications

Physical Form Crystalline Aggregates
Quantity 50 g
UN Number UN1477
Solubility Information Soluble in water.
Formula Weight 441.02 (350.94 Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Holmium(III) nitrate pentahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.