missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) oxide, 99.9%, (Total Rare Earth Oxides), -325 mesh

Product Code. 10694702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10694702 10 g Glass bottle 10g
10247023 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10694702 Supplier Thermo Scientific Acros Supplier No. 197140100
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
MDL Number MFCD00011053
InChI Key OWCYYNSBGXMRQN-UHFFFAOYSA-N
IUPAC Name diholmium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ho+3].[Ho+3]

Specifications

Boiling Point 3900.0°C
Melting Point 2330.0°C
Color Beige to Orange-Yellow
Physical Form Powder
Quantity 10 g
Packaging Glass bottle
Assay Percent Range 99.85% min. (Ho2O3) TREO base
Linear Formula HO2O3
Merck Index 15, 4765
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inorganic acids
Formula Weight 377.85
Percent Purity 99.9%
Chemical Name or Material Holmium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.