missing translation for 'onlineSavingsMsg'
Learn More

Holmium(III) Oxide, TRC

Product Code. 30137214
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137214 50 g 50g
30132545 5 g 5g
30012263 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137214 Supplier Toronto Research Chemicals Supplier No. TRCH66963350G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12055-62-8
Molecular Formula Ho2O3
Molecular Weight (g/mol) 377.86
Synonym Holmium oxide (Ho2O3), Diholmium trioxide, Holmia, Holmium oxide, Holmium sesquioxide, Holmium trioxide, Holmium(3+) oxide, Holmium(III) oxide, Nanotek WPC, Holmium(III) oxide, Holmia, Rare earth, Holmium(+3)oxide;
IUPAC Name oxo(oxoholmiooxy)holmium
SMILES O=[Ho]O[Ho]=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/2Ho.3O
Formula Weight 377.845
Chemical Name or Material Holmium oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.