missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homo-PROTAC cereblon degrader 1, MedChemExpress
Description
Homo-PROTAC cereblon degrader 1 (compound 15a) is a highly potent and efficient Cereblon (CRBN) degrader with only minimal effects on IKZF1 and IKZF3.
Specifications
Specifications
| Chemical Name or Material | Homo-PROTAC cereblon degrader 1 |
| CAS | 2244520-98-5 |
| Color | Green |
| Purity Grade Notes | Research |
| Molecular Formula | C32H32N6O10 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 200 mg/mL (302.74 mM; Need ultrasonic) |
| SMILES | O=C(N1C2CCC(NC2=O)=O)C3=CC=CC(NCCOCCOCCNC4=C5C(C(N(C6CCC(NC6=O)=O)C5=O)=O)=CC=C4)=C3C1=O |
| Molecular Weight (g/mol) | 660.63 |
| Formula Weight | 660.63 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?