missing translation for 'onlineSavingsMsg'
Learn More

Homomorpholine hydrochloride, 98%

Product Code. 11470367
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470367 5 g 5g
11460367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470367 Supplier Thermo Scientific Alfa Aesar Supplier No. B21749.06
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 178312-62-4 | C5H12ClNO | 137.607 g/mol

Homomorpholine Hydrochloride is a useful synthetic intermediate. It is used to prepare homomorpholine oxazolidinone with antibacterial activities. It is also used to synthesize A2A adenosine receptor antagonists for the treatment of Parkinson's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 178312-62-4
Molecular Formula C5H12ClNO
Molecular Weight (g/mol) 137.607
MDL Number MFCD02683070
InChI Key LJRCWNIWOVZLKS-UHFFFAOYSA-N
Synonym 1,4-oxazepane hydrochloride, homomorpholine hydrochloride, 1,4-oxazepine, hexahydro-, hydrochloride, homomorpholine hcl, 1,4-oxazepan hydrochloride, 1,4 oxazepane hydrochloride, hexahydro-1,4-oxazepine hydrochloride, homomorpholine-hydrochloride, homo-morpholine hydrochloride, homomorph-oline hydrochloride
PubChem CID 23090214
IUPAC Name 1,4-oxazepane;hydrochloride
SMILES C1CNCCOC1.Cl

Specifications

Melting Point 158°C to 164°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 137.61
Percent Purity 98%
Chemical Name or Material Homomorpholine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.