missing translation for 'onlineSavingsMsg'
Learn More

Homophthalic anhydride, 97+%

Product Code. 11452967
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11452967

Brand: Thermo Scientific Alfa Aesar A15538.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 703-59-3 | C9H6O3 | 162.144 g/mol

It is used as a chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a chemical and organic intermediates.

Solubility
Decomposes in water.

Notes
Moisture sensitive. Storage temperature 15 - 30°C. Store under inert gas. Store away from exposure to moisture, strong oxidizing agents, strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-59-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.144
MDL Number MFCD00006894
InChI Key AKHSBAVQPIRVAG-UHFFFAOYSA-N
Synonym homophthalic anhydride, 1,3-isochromandione, isochroman-1,3-dione, 1h-2-benzopyran-1,3 4h-dione, homophthalic acid anhydride, 4h-2-benzopyran-1,3-dione, 1h-isochromene-1,3 4h-dione, 3,4-dihydro-1h-2-benzopyran-1,3-dione, 4h-benzo c pyran-1,3-dione, homophthalic anhydride 2-carboxyphenylacetic anhydride
PubChem CID 12801
IUPAC Name 4H-isochromene-1,3-dione
SMILES C1C2=CC=CC=C2C(=O)OC1=O

Specifications

Melting Point 140°C to 144°C
Quantity 10 g
Beilstein 5244
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 162.14
Percent Purity ≥97%
Chemical Name or Material Homophthalic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.