missing translation for 'onlineSavingsMsg'
Learn More

Homosalate-d4, TRC

Product Code. 30396404
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396404 10 mg 10mg
30400898 1 mg 1mg
30397571 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396404 Supplier Toronto Research Chemicals Supplier No. TRCH61400210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Homosalate-d4
Purity Grade Notes HPLC
Molecular Formula C16 D4 H18 O3
Quantity 10 mg
Synonym Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester-d4, 3,3,5-Trimethylcyclohexanol Salicylate-d4, 3,3,5-Trimethylcyclohexyl Salicylate-d4, Eusolex HMS-d4, Filtrosol A-d4, Heliopan-d4, Heliopan HMS-d4, Heliophan-d4, Homomenthyl Salicylate-d4, Kemester-d4, Kemester HMS-d4, NSC 164918-d4, Neo Heliopan-d4, Neo Heliopan HMS-d4, Parsol HMS-d4, Uniderm Homsal-d4
InChI Formula InChI=1S/C16H22O3/c1-11-8-12(10-16(2,3)9-11)19-15(18)13-6-4-5-7-14(13)17/h4-7,11-12,17H,8-10H2,1-3H3/i4D,5D,6D,7D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OC2CC(C)CC(C)(C)C2)c(O)c1[2H]
IUPAC Name (3,3,5-trimethylcyclohexyl) 2,3,4,5-tetradeuterio-6-hydroxy-benzoate
Molecular Weight (g/mol) 266.37
Formula Weight 266.182
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.