missing translation for 'onlineSavingsMsg'
Learn More

HONB, TRC

Product Code. 30403531
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403531 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403531 Supplier Toronto Research Chemicals Supplier No. TRCH66955050G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HONB
CAS 21715-90-2
Molecular Formula C9H9NO3
Quantity 50 g
Synonym 3a,4,7,7a-Tetrahydro-2-hydroxy-4,7-methano-1H-isoindole-1,3(2H)-dione, N-Hydroxy-5-norbornene-2,3-dicarboximide, 3,5-Dioxo-4-azatricyclo[5.2.1.02'6]dec-8-en-4-ol, HONB, N-Hydroxy-5-bicyclo[2.2.1]heptene-2,3-dicarboximide, N-Hydroxy-5-norbornene-2,3-dicarboximide, NSC 100740, NSC 12953
InChI Formula InChI=1S/C9H9NO3/c11-8-6-4-1-2-5(3-4)7(6)9(12)10(8)13/h1-2,4-7,13H,3H2/t4-,5+,6?,7?
SMILES ON1C(=O)C2[C@@H]3C[C@@H](C=C3)C2C1=O
Molecular Weight (g/mol) 179.17
Formula Weight 179.0582
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.