missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Elano S3 Loafer Shoes

Product Code. 15221044
Change view
Click to view available options
Size:
42
43
44
45
Unit Size:
1 pair
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15221044 42 1 pair
15241044 44 1 pair
15231044 43 1 pair
15251044 45 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15221044 Supplier Honeywell™ Supplier No. 654301542/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance EN ISO 20345:2011, CI and SRC Certified
Color Black
Footbed Material Leather
For Use With (Application) For professional use such as army - defence, fishing, textile industries, transportation
Material Leather
Size 42
Sole Features Perforated Leather insole, Non-metallic antipuncture midsole, protects 100% of the foot surface
Sole Material Double density Polyurethane
Style Loafer
Resistant To Water resistant
Waterproof Yes
Product Type Loafer Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.