missing translation for 'onlineSavingsMsg'
Learn More

Sperian™ Honeywell™ Laser Lite Single-Use Uncorded Earplugs

Product Code. 11846940
Change view
Click to view available options
Shape: T-shape
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11846940 Shape: T-shape
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11846940 Supplier Sperian™ Supplier No. 3301105
  • ONLINE EXCLUSIVE PRICE
    97.29 €
    44.18 € / 200 pairs
    Save 53.11 €
    54% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Makes protection fun and highly visible. Sperian™ Honeywell™ Laser Lite Single-Use Uncorded Earplugs are made from self-adjusting polyurethane foam which fits virtually every wearer.

  • Vibrant colors
  • Contoured t-shape aids in easy insertion and removal from ear canal
  • Comfortable long-term wear
  • Ideal for process industries with low tolerances for packaging waste
TRUSTED_SUSTAINABILITY

Specifications

Color Magenta, Yellow
Certifications/Compliance EN 352-2:1993 / EN-24869-1:1993, ISO 9001/2000
Size Universal
For Use With (Application) For process industries with low tolerances
Quantity 200/Pk.
Plug Shape T-Shape
Product Type Ear Plugs
Disposable Single-use
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.