missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ Metal Hearing Protection
Description
- Designed to fit and seal to the workers ear perfectly
- Reduces background noises to safe levels
- Efficiently collect noise exposure, helping identify risks and take action
Specifications
Specifications
| Color | Black |
| Certifications/Compliance | ISO 9001/2000 |
| For Use With (Application) | Noise Blocking Ear Muff and For Construction Workers |
| Product Type | Hearing Protection |
| Material | Metal |
| Style | Helmet-mounted |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?