missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Pro'Clean S2 Shoes

Product Code. 15289503
Change view
Click to view available options
Size:
35
36
38
39
41
44
Unit Size:
1 pair
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15289503 41 1 pair
15239503 36 1 pair
15229503 35 1 pair
15249503 38 1 pair
15269503 39 1 pair
15219513 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15289503 Supplier Honeywell™ Supplier No. 624612741/7

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Certifications/Compliance EN ISO 20345: 2011, SRC Certified
Color White
For Use With (Application) For professional use such as catering, industrial cleaning, laboratory, medical and pharmaceutical
Material Microfiber
Size 41
Sole Features Removable, anti-static insole
Sole Material Double density Polyurethane
Toe Material Spacium non-metallic
Resistant To Abrasion, fuel oil and grease
Waterproof Yes
Product Type Shoes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.