missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Supervizor Face Shields

Product Code. 11836990
Change view
Click to view available options
Model:
SB600 and SV9PC
SV9AC
SV9PC
Description:
200 mm Clear Acetate Visor
230 mm Clear Polycarbonate Visor
Browguard with Ratchet Headband
Browguard with Ratchet Headband and 230mm Clear Polycarbonate Visor
Lens Material:
Acetate
Polycarbonate
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Description Lens Material Unit Size
11836990 SV9AC 200 mm Clear Acetate Visor Acetate Each
11826990 SV9PC 230 mm Clear Polycarbonate Visor Polycarbonate Each
12990395 SB600 and SV9PC Browguard with Ratchet Headband and 230mm Clear Polycarbonate Visor Polycarbonate Each
11816990 - Browguard with Ratchet Headband Polycarbonate Each
15670324 - Browguard with Ratchet Headband Polycarbonate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11836990 Supplier Honeywell™ Supplier No. 1002307
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Robust, heavy-duty, helmet-mounted face shield range with hearing protection compatibility. Sperian™ Honeywell™ Supervizor Face Shields have a wide visor selection that can be tailored to the specific hazard/application.

  • Recommended for use in the chemical and forestry industries, among others
  • Protects against hazards such as abrasion, airborne particles, biohazards, chemicals, gases, vapors, and smoke
  • Maximum protection, allowing full unrestricted head movement
  • Creased visor eliminates gaps and visor profile follows contours of the face
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 166 1F
Width (Metric) 200 mm
Description 200 mm Clear Acetate Visor
Product Type Supervisor Faceshield
Model SV9AC
No. per Pack 1 Ea.
Lens Material Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.