missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ ThermoSafe™ Compact Insulated Shipper

Product Code. 11756028 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W x H):
6 x 6 x 6in.
8 x 8 x 11in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11756028 6 x 6 x 6in. Each
11775509 8 x 8 x 11in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11756028 Supplier Cole-Parmer™ Supplier No. 0373020
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose from a variety of sizes to meet your refrigeration needs with these compact insulated shippers. Cole Parmer™ ThermoSafe™ Compact Insulated Shippers are ideal for shipping non-hazardous materials.

  • Compact shippers have a cylindrical interior to hold tubes, vials and other small items
  • Add as many ice packs as you need
  • For non-hazardous materials only

Specifications

Height (English) 6 in.
Length (English) 6 in.
Width (English) 6 in.
Dimensions (L x W x H) 6 x 6 x 6in.
Diameter (English) 3 in.
Product Type Compact Insulated Shipper
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.