missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, 98+%, Thermo Scientific™

Induces apoptosis in human squamous lung cancer cells

116.00 € - 557.00 €

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18H18O2
Molecular Weight (g/mol) 266.34
MDL Number MFCD00016674
InChI Key FVYXIJYOAGAUQK-UHFFFAOYSA-N
Synonym honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
PubChem CID 72303
ChEBI CHEBI:5759
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15445679
View Documents
Thermo Scientific Alfa Aesar
J63434.MA
10mg
116.00 €
10mg
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15455679
View Documents
Thermo Scientific Alfa Aesar
J63434.MB
25mg
211.00 €
25mg
View alternative products
Save Up to 
»
Estimated Shipment: 06-08-2026
Add to basket
 
 Restricted Item
15465679
View Documents
Thermo Scientific Alfa Aesar
J63434.MC
100mg
557.00 €
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 06-08-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

35354-74-6
266.34
FVYXIJYOAGAUQK-UHFFFAOYSA-N
72303
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
C18H18O2
MFCD00016674
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
CHEBI:5759
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

35354-74-6
MFCD00016674
14,4742
FVYXIJYOAGAUQK-UHFFFAOYSA-N
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
72303
266.34
Honokiol
C18H18O2
UN3077
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C
266.34
CHEBI:5759
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.