missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, 98+%, Thermo Scientific™

Induces apoptosis in human squamous lung cancer cells

Brand:  Thermo Scientific Alfa Aesar J63434.MB

Quantity 25mg

 View more versions of this product

Product Code. 15455679

  • 211.00 € / 25mg
 Restricted Item
Estimated Shipment: 06-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

35354-74-6
266.34
FVYXIJYOAGAUQK-UHFFFAOYSA-N
72303
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
C18H18O2
MFCD00016674
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
CHEBI:5759
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C

Specifications

35354-74-6
MFCD00016674
UN3077
honokiol, 5,3'-diallyl-2,4'-dihydroxybiphenyl, 3,5'-diallyl-4,2'-dihydroxybiphenyl, unii-11513cco0n, chembl16901, 3',5-diallyl-1,1'-biphenyl-2,4'-diol, 4-allyl-2-3-allyl-4-hydroxy-phenyl phenol, 3',5-diallyl-2,4'-biphenyldiol, 3',5-diallylbiphenyl-2,4'-diol, 5,3′-diallyl-2,4′-dihydroxydiphenyl
C=CCC1=CC(=C(C=C1)O)C2=CC(=C(C=C2)O)CC=C
266.34
CHEBI:5759
≥98%
C18H18O2
25mg
14,4742
FVYXIJYOAGAUQK-UHFFFAOYSA-N
2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
72303
266.34
Honokiol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.