Learn More
Medicom™ Hopen™ Single Use Hygiene Gown
Medicom™ Hopen™ Single Use Hygiene Gown without pockets, is microporous and waterproof
Brand:  Medicom™ 40.550/04
 View more versions of this product
Product Code. 16544081
  • 157.00 € / Pack of 50
Only null left
Estimated Shipment: 23-04-2026
Add to basket
Description
Description
  • Reduces contamination of the operator, but also sensitive products and processes in a protected atmosphere
  • Protects against radioactive particles and infectious agents
  • Microporous, Anti-Static Hygiene Gown is comfortable and light
  • Polypropylene gown lined with laminated polyethylene for a waterproof, hydrophobic seal
  • Available in mulitple sizes, and closure types
Specifications
Specifications
X-Large
Polypropylene
Press studs
Yes
Radioactive Particles, Infectious Agents
Microporous, Waterproof
Manufactured under an ISO 9001 Certified Quality System
Category 1 Personal Protective Equipment
Unisex
White
Elastic
Non-sterile
None
Sewn
EN 1149-5
Individual, Bag