Learn More
Medicom™ Hopen™ Pocketed Hygiene Gown
GREENER_CHOICE
Medicom™ Hopen™ Pocketed Hygiene Gown with press-stud fasteners, available in two colors
Brand:  Medicom™ 40.401/05
Product Code. 16554061
  • 52.45 € / Pack of 50
Only null left
Estimated Shipment: 02-03-2026
Add to basket
Description
Description
  • Gown protects the operator, as well as sensitive products/processes that could be contaminated in a protected atmosphere
  • Protects user against radioactive particles, and infectious agents
  • Reduces the risk of soiling related to the user's activity
  • Pleasant and light, polypropylene fabric is breathable yet durable
  • Variety of colors allows for possibility of identification by color codes
  • Two pockets: one on the chest, and one on the lower thigh
  • Individually packaged for controlled hygiene, and distribution
  • Pockets located on the upper left, and lower right of the gown